

Hydrochloric acid 37%, extra pure
Catalog # ACRO450560050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 L
- Seq:5190
Specifications
Specification Test Results
- Appearance:Clear liquid
- Color scale:≤20 APHA
- Residue after evaporation:≤0.005 %
- Titration with NaOH:35.0 to 39.0 %
Specification Test Results
About this item
:
Boiling Pt:57 °C (760 mmHg)
Melting Pt:–35 °C
Density:1.18 g/cm³
Storage Temperature:Ambient
ADR:8,II
:36.46 g/mol
UN:1789
:GHS05
:GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324