
3'-Chloro-4'-hydroxyacetanilide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3964-54-3
:
Formula:C₈H₈ClNO₂
MDL Number:MFCD01318123
:185.6100 g/mol
Melting Pt:144…146 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Off-white to light pink to light brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:143 to 146 °C
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together