1-Acetylpyrene 97%
Catalog # ACRO449300050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Yellow to light brown Crystalline powder or flakes
- Infrared spectrum:Conforms
- Melting point:86 to 89 °C
- HPLC:≥96.0 %
Specification Test Results
About this item
:
:244.29 g/mol
Melting Pt:86…89 °C
Storage Temperature:Ambient
CAS Number:3264-21-9
Formula:C₁₈H₁₂O
MDL Number:MFCD00021663