

1-Methoxy-2,2,2-trifluoroethanol 90%, Technical Grade
Catalog # ACRO449130250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3260 to 1.3290 (20°C, 589 nm)
- GC:≥88 %
Specification Test Results
About this item
:
Boiling Pt:104…106 °C
Density:1.36 g/cm³
Flash Pt:42 °C
Storage Temperature:Ambient
ADR:3,III
:130.07 g/mol
UN:1993
:GHS02
:GHS07
CAS Number:431-46-9
Formula:C₃H₅F₃O₂
MDL Number:MFCD00013572