Order Entry
Czech Republic
ContactUsLinkComponent
 
 
4-Bromo-2,6-dinitrotoluene 97%
 
undefined
 
4-Bromo-2,6-dinitrotoluene 97%

 

  • Appearance:
    White to yellow Crystalline solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    86 to 89 °C
  • GC:
    ≥96.0 %

 

 

Frequently Bought Together