
5-Methoxyisatin 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:39755-95-8
:
Formula:C₉H₇NO₃
MDL Number:MFCD00169023
:177.16 g/mol
Melting Pt:201…203 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark red to brown Powder
- Infrared spectrum:Conforms
- Melting point:199 to 202 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together