



Sodium borohydride 99%, granules, VenPure™ SF, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16940-66-2
:
Formula:NaBH₄
MDL Number:MFCD00003518
:37.83 g/mol
Melting Pt:360 °C (dec.)
Density:1,07 g/cm³ (20 °C)
Flash Pt:69 °C
UN:1426
ADR:4.3,I
:GHS06
:GHS05
:GHS02
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Granules
- Assay:≥98.5 % (Hydrogen evolution)
- Iron (Fe):≤5 ppm
- Silicon dioxide (SiO2):21 to 500 ppm
- Fines:≤8.5 % (< 0.5 cm)
Specification Test Results
Frequently Bought Together