
Picolinamidine hydrochloride 97%
Catalog # ACRO448610050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to off-white Solid
- Infrared spectrum:Conforms
- Titration Argentometric:≥96.0 to ≤104.0 %
- HPLC:≥96.0 %
Specification Test Results
About this item
:
:157.60 g/mol
Melting Pt:146…148 °C
:GHS07
Storage Temperature:Ambient
CAS Number:51285-26-8
Formula:C₆H₈ClN₃
MDL Number:MFCD00052271