Silica gel humidity indicator cards cobalt-free
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:63231-67-4
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
Storage Temperature:Ambient
Specification Test Results
- Suitability:Passes test
Specification Test Results
Frequently Bought Together