

N,N-Diisopropylethylenediamine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:121-05-1
:
Formula:C₈H₂₀N₂
MDL Number:MFCD00015015
:144.26 g/mol
Boiling Pt:170 °C (760 mmHg)
Density:0.83 g/cm³
Flash Pt:54 °C
Storage Temperature:Ambient
UN:2734
ADR:8,II
:GHS02
:GHS05
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4420 to 1.4440 (20°C, 589 nm)
- Titration with HClO4:≥96.0 %
Specification Test Results
Frequently Bought Together