
N-(tert-Butoxycarbonyl)-5-aminovaleric acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:27219-07-4
:
Formula:C₁₀H₁₉NO₄
MDL Number:MFCD00076903
:217.27 g/mol
Boiling Pt:160…168 °C
Melting Pt:45…52 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:48 to 54 °C
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together