Order Entry
Czech Republic
ContactUsLinkComponent
Sodium dihydrogen phosphate, anhydrous 98%, extra pure
Sodium dihydrogen phosphate, anhydrous 98%, extra pure
Certificates
undefined
Sodium dihydrogen phosphate, anhydrous 98%, extra pure
Certificates

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Water:
    ≤2 %
  • Loss on drying:
    ≤1 % (130°C)
  • pH:
    4.1 to 4.5
  • Titration with HClO4:
    97.5 to 103 %
  • Heavy metals (as Pb):
    ≤0.002 %

Specification Test Results

Related Information

Frequently Bought Together