Disodium hydrogen phosphate, anhydrous 98+%, extra pure
:7558-79-4
:
:Na₂HPO₄
:MFCD00003496
:141.96 g/mol
:250 °C
:1,53 g/cm³ (20 °C)
:Ambient
- Appearance:White Powder
- Loss on drying:≤5 % (130°C)
- pH:8.7 to 9.4 (5 % in water at 25°C)
- Titration with HClO4:98.0 to 102.0 % (On dried substance)
- Sodium dihydrogenphosphate:≤2.5 %
- Heavy metals (as Pb):≤20 ppm
Frequently Bought Together