Order Entry
Czech Republic
ContactUsLinkComponent
Sodium dihydrogen phosphate, anhydrous 99% for biochemistry
Sodium dihydrogen phosphate, anhydrous 99% for biochemistry
Certificates
undefined
Sodium dihydrogen phosphate, anhydrous 99% for biochemistry

Specification Test Results

  • Appearance:
    White Crystalline powder and/or chunks
  • Loss on drying:
    ≤1 % (130°C)
  • Solubility:
    (1 M in water) Clear colorless
  • UV:
    (1 M in water)
  • pH:
    4.0 to 6.0 (1 M in water)
  • Titration with HClO4:
    ≥98.5 %
  • Heavy metals (as Pb):
    ≤0.001 %
  • Insoluble matter:
    ≤0.2 % (in water)

Specification Test Results

Related Information

Frequently Bought Together