
Bismuth(III) bromide 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7787-58-8
:
Formula:BiBr₃
MDL Number:MFCD00014195
:448.69 g/mol
Boiling Pt:278 °C (11 mmHg)
Melting Pt:218 °C
Density:5,72 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3260
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Light yellow to yellow or light grey Crystalline powder or crystals
- Melting point:218 to 220°C
- Assay:96 to 103 % (Bi)
- Chloride (Cl):≤0.1 %
- Iron (Fe):≤0.05 %
Specification Test Results
Frequently Bought Together