Order Entry
Czech Republic
ContactUsLinkComponent
 
Magnesium nitrate hexahydrate, extra pure
 
undefined
Magnesium nitrate hexahydrate, extra pure

 

  • Appearance:
    White Crystals
  • Titration Complexometric:
    98 to 102 %
  • pH:
    5 to 8.2 (5 % in water)
  • Chloride (Cl):
    ≤0.001 %
  • Sulfate (SO4):
    ≤0.005 %
  • Iron (Fe):
    ≤0.001 %
  • Heavy metals (as Pb):
    ≤0.001 %

 

 

Frequently Bought Together