Order Entry
Czech Republic
ContactUsLinkComponent
L(+)-Potassium sodium tartrate tetrahydrate ACS
L(+)-Potassium sodium tartrate tetrahydrate ACS
Certificates
undefined
L(+)-Potassium sodium tartrate tetrahydrate ACS

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • pH:
    6.0 to 8.5 (5% soln. at 25°C)
  • Titration with HClO4:
    99.0 to 102.0 %
  • Chloride (Cl):
    ≤0.001 %
  • Phosphate (PO4):
    ≤0.002 %
  • Sulfate (SO4):
    ≤0.005 %
  • Ammonium (NH4):
    ≤0.002 %
  • Calcium (Ca):
    ≤0.005 %
  • Iron (Fe):
    ≤0.001 %
  • Heavy metals (as Pb):
    ≤5 ppm
  • Insoluble matter:
    ≤0.005 %

Specification Test Results

Related Information

Frequently Bought Together