Order Entry
Czech Republic
ContactUsLinkComponent
Magnesium sulphate heptahydrate 99% for biochemistry
Magnesium sulphate heptahydrate 99% for biochemistry
Certificates
undefined
Magnesium sulphate heptahydrate 99% for biochemistry

Specification Test Results

  • Appearance:
    White Crystals
  • Titration Complexometric:
    ≥98.5 %
  • Solubility:
    (1 M in water) Clear colorless
  • pH:
    5 to 8
  • Heavy metals (as Pb):
    ≤5 ppm

Specification Test Results

Related Information

Frequently Bought Together