Order Entry
Czech Republic
ContactUsLinkComponent
Diammonium hydrogen orthophosphate 99% for biochemistry
Diammonium hydrogen orthophosphate 99% for biochemistry
Certificates
undefined
Diammonium hydrogen orthophosphate 99% for biochemistry

Specification Test Results

  • Appearance:
    White Crystals
  • Solubility:
    (1 M in water) Clear colorless
  • UV:
    (1 M in water)
  • pH:
    7.5 to 9.0 (1 M in water)
  • Titration with HCl:
    ≥98.5 %
  • Heavy metals (as Pb):
    ≤5 ppm

Specification Test Results

Related Information

Frequently Bought Together