

(S)-2-(Trifluoromethyl)piperidine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:154727-51-2
:
Formula:C₆H₁₀F₃N
MDL Number:MFCD11226597
:153.15 g/mol
Boiling Pt:80 °C (200Torr)
UN:2733
ADR:3,III
:GHS02
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light brown Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
- NMR (1H-NMR):Consistent with structure
- Specific optical rotation:-10° to -20° (20°C, 589 nm) (c=4 in methanol)
Specification Test Results
Frequently Bought Together