
4,4,4,4',4',4'-Hexafluoro-DL-valine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16063-80-2
:
Formula:C₅H₅F₆NO₂
MDL Number:MFCD00014349
:225.09 g/mol
Melting Pt:207…211 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Powder or solid
- Infrared spectrum:Conforms
- Assay:96 to 104 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
Frequently Bought Together