
Sodium thiocyanate 98+% for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:540-72-7
:
Formula:NaSCN
MDL Number:MFCD00011123
:81.07 g/mol
Melting Pt:287 °C
Density:1,735 g/cm³ (20 °C)
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Titration Argentometric:≥98.0 %
- Chloride (Cl):≤0.01 %
- Sulfide (S):≤0.001 %
- Sulfate (SO4):≤0.01 %
- Sodium carbonate (Na2CO3):≤0.2 %
- Ammonium (NH4):≤0.002 %
- Iron (Fe):≤0.0002 %
- Heavy metals (as Pb):≤5 ppm
- Insoluble matter:≤0.005 % (in water)
Specification Test Results
Frequently Bought Together