Cyclohexane ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-82-7
:
Formula:C₆H₁₂
MDL Number:MFCD00003814
:84.16 g/mol
Boiling Pt:81 °C (760 mmHg)
Melting Pt:6.5 °C
Density:0.77 g/cm³
Flash Pt:–18 °C
UN:1145
ADR:3,II
:GHS07
:GHS08
:GHS09
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.02 %
- GC:≥99.0 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.002 %
- Substance darkened by H2SO4:Passes test
Specification Test Results
Frequently Bought Together