

Triethylsilane 99%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:617-86-7
:
Formula:C₆H₁₆Si
MDL Number:MFCD00009018
:116.28 g/mol
Boiling Pt:107…108 °C (760 mmHg)
Melting Pt:-156.9 °C
Density:0.72 g/cm³
Flash Pt:–3 °C
Storage Temperature:Ambient
UN:1993
ADR:3,II
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4100 to 1.4120 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Related Information
Frequently Bought Together