Methylmagnesium bromide 1 M in CPME (cyclopentyl methyl ether), AcroSeal®
:75-16-1
:
:CH₃BrMg
:MFCD00000041
:119.24 g/mol
:–1 °C
:3399
:4.3,I
:GHS02
:GHS05
:GHS07
:Ambient
- Appearance:Dark grey Solution (May develop some turbidity or precipitate)
- Titration Grignard:0.95 to 1.10 M
Frequently Bought Together