


Vinylmagnesium bromide 1 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1826-67-1
:
Formula:C₂H₃BrMg
MDL Number:MFCD00000042
:131.26 g/mol
Density:0.965 g/cm³
Flash Pt:–11 °C
UN:3399
ADR:4.3,I
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to red to brown Solution (May develop some turbidity or precipitate)
- Titration Active base:0.80 to 1.10 M
Specification Test Results
Related Information
Frequently Bought Together