
3-(Tetrahydropyran-2-yloxy)prop-1-en-1-ylboronic acid pinacol ester 90%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:642066-70-4
:
Formula:C₁₄H₂₅BO₄
MDL Number:MFCD03788736
:268.16 g/mol
Boiling Pt:79 °C (,1Torr)
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless Oily liquid
- GC:≥88.0 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
Frequently Bought Together