Order Entry
Czech Republic
ContactUsLinkComponent
Tetrachloroethylene 99+% for HPLC
Irritant
Health
Environment
Tetrachloroethylene 99+% for HPLC
Certificates
undefined
Irritant
Health
Environment
Tetrachloroethylene 99+% for HPLC

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.05 %
  • GC:
    ≥99.0 %
  • UV:
    at 290 nm A: ≤1
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.0005 %

Specification Test Results

Related Information

Frequently Bought Together