Order Entry
Czech Republic
ContactUsLinkComponent
Norfloxacin, 98%, analytical standard
Norfloxacin, 98%, analytical standard
  ACRO445260050
 :  
Norfloxacin, 98%, analytical standard
  ACRO445260050
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • CAS No.:
    70458-96-7
  • Description:
    Norfloxacin 98%
  • Melting point:
    218…226 °C
  • Formula:
    C₁₆H₁₈FN₃O₃
  • Description:
    Norfloxacin