
5-(Methoxycarbonyl)-2-methylphenylboronic acid pinacol ester 95%
Catalog # ACRO445110010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to off-white to brown Solid
- Infrared spectrum:Conforms
- Melting point:157 to 162 °C
- GC:≥94.0 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
About this item
:
:276.136 g/mol
Melting Pt:157…159 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:882679-40-5
Formula:C₁₅H₂₁BO₄
MDL Number:MFCD11520530