Order Entry
Czech Republic
ContactUsLinkComponent
1-Isopropyl-1H-pyrazole-4-boronic acid pinacol ester 97%
Irritant
1-Isopropyl-1H-pyrazole-4-boronic acid pinacol ester 97%
Certificates
undefined
Irritant
1-Isopropyl-1H-pyrazole-4-boronic acid pinacol ester 97%

Specification Test Results

  • Appearance:
    White to off-white to light brown Solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    38 to 46 °C
  • GC:
    ≥96.0 %
  • NMR (1H-NMR):
    Consistent with structure

Specification Test Results

Related Information

Frequently Bought Together