

Hydrochloric acid 36%, Technical Grade
Katalogové číslo ACRO444390250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic drum
- Pk:25 L
- Seq:5327
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Color scale:≤20 APHA
- Titration with NaOH:35 to 37 %
Specification Test Results
About this item
:
Boiling Pt:57 °C (760 mmHg)
Melting Pt:–35 °C
Density:1.16 g/cm³
Storage Temperature:Ambient
ADR:8,II
:36.46 g/mol
UN:1789
:GHS05
:GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324