
Sodium hydroxide 30% in water, Technical Grade
Catalog # ACRO444350050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic drum
- Pk:5 L
- Seq:5190
Specifications
Specification Test Results
- Appearance:Clear colorless Solution
- Titration with HCl:28.0 to 33.5 %
Specification Test Results
About this item
:
:40 g/mol
Boiling Pt:120 °C (760 mmHg)
Melting Pt:8 °C
Density:1.31 g/cm³
Storage Temperature:Ambient
UN:1824
ADR:8,II
:GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548