Order Entry
Czech Republic
ContactUsLinkComponent
Water, high density, demineralised
Water, high density, demineralised
Catalog # ACRO444300250
   Safer Human and Environmental Health
CAS Number:  
undefined
Water, high density, demineralised
Catalog # ACRO444300250
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Plastic can
  • Pk:
    25 L
  • Seq:
    5327
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • pH:
    6.0 to 8.0
  • Conductivity:
    ≤15 µS/cm

Specification Test Results

About this item

 :
 :18.02 g/mol
Boiling Pt:100 °C (760 mmHg)
Melting Pt:0 °C
Density:1 g/cm³
Storage Temperature:Ambient
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332