
(R)-4-Hydroxypyrrolidin-2-one 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:22677-21-0
:
Formula:C₄H₇NO₂
MDL Number:MFCD00273364
:101.11 g/mol
Melting Pt:155…156 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
- Specific optical rotation:+54° to +60° (20°C, 589 nm) (c=0.36 in water)
Specification Test Results
Frequently Bought Together