n-Pentane, extra dry over Molecular Sieve 99+%, AcroSeal®
:109-66-0
:
:H₃C(CH₂)₃CH₃
:MFCD00009498
:72.15 g/mol
:35…36 °C (1013 hPa)
:–130 °C
:0.626 g/cm³
:–49 °C
:1265
:3,II
:GHS07
:GHS08
:GHS09
:GHS02
:Ambient
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥99.0 %
Frequently Bought Together