ContactUsLinkComponent
(S)-(+)-3-Fluoropyrrolidine hydrochloride 97%
Irritant
(S)-(+)-3-Fluoropyrrolidine hydrochloride 97%
Certificates
undefined
Irritant
(S)-(+)-3-Fluoropyrrolidine hydrochloride 97%
Specification Test Results
  • Appearance:
    White to light brown Powder
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    ≥96.0 to ≤104.0 %
  • Specific optical rotation:
    +5° to +11° (20°C, 589 nm) (c=1 in MeOH)