Order Entry
Czech Republic
ContactUsLinkComponent
5-Chloro-2-fluorobenzoic acid 97%
Irritant
5-Chloro-2-fluorobenzoic acid 97%
Catalog # ACRO442510050
CAS Number:  
undefined
Irritant
5-Chloro-2-fluorobenzoic acid 97%
Catalog # ACRO442510050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    151 to 157 °C
  • HPLC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :174.56 g/mol
Melting Pt:151…153 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:394-30-9
Formula:C₇H₄ClFO₂
MDL Number:MFCD00665762