Order Entry
Czech Republic
ContactUsLinkComponent
4-Fluoro-2-methylbenzenesulphonyl chloride 97%
Corrosive
4-Fluoro-2-methylbenzenesulphonyl chloride 97%
Catalog # ACRO442370050
CAS Number:  
undefined
Corrosive
4-Fluoro-2-methylbenzenesulphonyl chloride 97%
Catalog # ACRO442370050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :208.64 g/mol
Boiling Pt:150…152 °C (92Torr)
Density:1,433 g/cm³ (20 °C)
Flash Pt:110 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
 :GHS05
CAS Number:7079-48-3
Formula:C₇H₆ClFO₂S
MDL Number:MFCD03094390