Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
 
2-Bromo-2',4'-difluoroacetophenone 95%
 
undefined
 
 
 
2-Bromo-2',4'-difluoroacetophenone 95%

 

  • Appearance:
    White to light yellow Low melting solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    30 to 35 °C
  • GC:
    ≥94.0 %

 

 

Frequently Bought Together