Order Entry
Czech Republic
ContactUsLinkComponent
2,3-Dimethoxy-5-methyl-p-benzoquinone 98%
Irritant
2,3-Dimethoxy-5-methyl-p-benzoquinone 98%
Certificates
undefined
Irritant
2,3-Dimethoxy-5-methyl-p-benzoquinone 98%

Specification Test Results

  • Appearance:
    Orange to red Crystalline powder or needles
  • Infrared spectrum:
    Conforms
  • Melting point:
    58 to 62 °C
  • HPLC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together