
2-Fluoro-6-(trifluoromethyl)benzyl bromide 95%
Catalog # ACRO442160050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to white to light yellow Crystalline powder or low melting solid
- Infrared spectrum:Conforms
- Melting point:38 to 45 °C
- GC:≥94.0 %
Specification Test Results
About this item
:
:257.03 g/mol
Boiling Pt:197 °C (1013 hPa)
Melting Pt:42…44 °C
Flash Pt:107 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
CAS Number:239087-08-2
Formula:C₈H₅BrF₄
MDL Number:MFCD00082477