Order Entry
Czech Republic
ContactUsLinkComponent
2,6-Dichloro-4-(trifluoromethoxy)aniline 96%
Irritant
2,6-Dichloro-4-(trifluoromethoxy)aniline 96%
Catalog # ACRO442300050
CAS Number:  
undefined
Irritant
2,6-Dichloro-4-(trifluoromethoxy)aniline 96%
Catalog # ACRO442300050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to white to light brown Crystalline needles or low melting solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    30 to 35 °C
  • GC:
    ≥95.0 %

Specification Test Results

About this item

 :
 :246.02 g/mol
Melting Pt:32…34 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:99479-66-0
Formula:C₇H₄Cl₂F₃NO
MDL Number:MFCD00190128