6-(Hydroxymethyl)benzothiazole 97%
ACRO441330010
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:White to yellow Crystalline powder or needles
- Infrared spectrum:Conforms
- Melting point:103 to 106 °C
- HPLC:≥96.0 %
:
:165.21 g/mol
:103…105 °C
:Ambient
:19989-66-3
:C₈H₇NOS
:MFCD04972625