
(R)-N-Fmoc-α-methylvaline 98%, 98% ee
Catalog # ACRO441145000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mg
- Seq:2725
Specifications
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Enantiomeric excess:≥98.0 %
- Specific optical rotation:-7.5° to -9.5° (20°C, 589 nm) (c=1 in CHCl3)
- Residual solvents:≤10 % MTBE
Specification Test Results
About this item
:
:353.418 g/mol
Melting Pt:37…53 °C
Storage Temperature:Refrigerator
UN:3175
ADR:4.1,II
:GHS02
CAS Number:616867-28-8
Formula:C₂₁H₂₃NO₄
MDL Number:MFCD12031696