
2-Methyl-L-cysteine hydrochloride 98%, 98% ee
Catalog # ACRO441101000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Elemental Analysis:27.29 to 28.69 % (C)
- Specific optical rotation:+6° to +10° (20°C, 589 nm) (c=1 in water)
Specification Test Results
About this item
:
Melting Pt:120 °C (dec.)
:171.64 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:148766-37-4
Formula:C₄H₁₀ClNO₂S
MDL Number:MFCD08063384