


Nonylmagnesium bromide 1 M in diethyl ether, AcroSeal®
Catalog # 440781000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Clear colorless to brown Liquid
- Titration Grignard:0.90 to 1.10 M
Specification Test Results
About this item
:
:231.46 g/mol
Boiling Pt:35 °C (760 mmHg)
Density:0.849 g/cm³
Flash Pt:–30 °C
Storage Temperature:Ambient
UN:2924
ADR:3,I
:GHS02
:GHS05
:GHS07
CAS Number:39691-62-8
Formula:C₉H₁₉BrMg
MDL Number:MFCD03094048