
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4568-71-2
:
Formula:C₁₃H₁₆ClNOS
MDL Number:MFCD00011959
Melting Pt:142…146 °C
:269.79 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:141 to 147 °C
- Elemental Analysis:56.52 to 59.32 % C
Specification Test Results
Frequently Bought Together