Order Entry
Czech Republic
ContactUsLinkComponent
N,N,N',N'-Tetramethyl-S-(1-oxido-2-pyridyl)thiouronium hexafluorophosphate 97%
Irritant
N,N,N',N'-Tetramethyl-S-(1-oxido-2-pyridyl)thiouronium hexafluorophosphate 97%
Catalog # ACRO440260050
CAS Number:  
undefined
Irritant
N,N,N',N'-Tetramethyl-S-(1-oxido-2-pyridyl)thiouronium hexafluorophosphate 97%
Catalog # ACRO440260050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to yellow Powder
  • Infrared spectrum:
    Conforms
  • Elemental Analysis:
    30.41 to 34.29 % (C)

Specification Test Results

About this item

 :
 :371.28 g/mol
Melting Pt:111.7…113.6 °C
 :GHS07
Storage Temperature:Freezer
CAS Number:212333-72-7
 :MFCD08274618
Formula:C₁₀H₁₆F₆N₃OPS
MDL Number:MFCD05865266