Order Entry
Czech Republic
ContactUsLinkComponent
O-(Benzotriazol-1-yl)-N,N,N',N'-bis(tetramethylene)uronium hexafluorophosphate 98%
Irritant
O-(Benzotriazol-1-yl)-N,N,N',N'-bis(tetramethylene)uronium hexafluorophosphate 98%
Catalog # ACRO440250250
CAS Number:  
undefined
Irritant
O-(Benzotriazol-1-yl)-N,N,N',N'-bis(tetramethylene)uronium hexafluorophosphate 98%
Catalog # ACRO440250250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Elemental Analysis:
    40.73 to 42.81 % (C)

Specification Test Results

About this item

 :
 :431.323 g/mol
Melting Pt:176 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:105379-24-6
Formula:C₁₅H₂₀F₆N₅OP
MDL Number:MFCD00191770